Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA

Catalog No. DYC869B5
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DYC869B5 5 96-well Plates
DYC869B2 2 96-well Plates
DYC869BE 15 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC869B5 Supplier R&D Systems Supplier No. DYC869B5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.

The Phospho-p38 alpha (T180/Y182) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat p38 alpha.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit (for 5 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Format 5 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 1432
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.