Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-p38 MAPK gamma/delta (Tyr185, Tyr182) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105907 Supplier Invitrogen™ Supplier No. PA5105907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of p38 gamma/delta only when phosphorylated at Tyr185/182.

Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. The protein encoded by this gene functions as a signal transducer during differentiation of myoblasts to myotubes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-p38 MAPK gamma/delta (Tyr185, Tyr182)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAPK13
Gene Accession No. O08911, O15264, P53778, Q63538, Q9WTY9, Q9Z1B7
Gene Alias AW123708; ERK3; ERK6; ERK-6; Exon skip; extracellular signal-regulated kinase 6; MAP kinase 12; MAP kinase 13; MAP kinase p38 delta; MAP kinase p38 gamma; MAPK 12; MAPK 13; MAPK12; MAPK13; MAPK-13; mitogen activated protein kinase 12; mitogen activated protein kinase 13; mitogen-activated protein kinase 12; Mitogen-activated protein kinase 13; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 delta; Mitogen-activated protein kinase p38 gamma; p38 delta MAP kinase; p38delta; P38GAMMA; PRKM12; Prkm13; SAP kinase-3; SAPK/Erk/kinase 4; SAPK3; SAPK-3; SAPK4; Serk4; stress activated protein kinase 3; stress-activated protein kinase 3; Stress-activated protein kinase 4
Gene Symbols MAPK12, MAPK13
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAPK12(Accession P53778), corresponding to amino acid residues around phosphorylated Tyr185.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26415, 29513, 29857, 5603, 60352, 6300
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.