Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-p41-ARCb (Thr21) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143652 Supplier Invitrogen™ Supplier No. PA5143652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 41 kDa* protein corresponding to the molecular mass of Arpc1b on SDS-PAGE immunoblots of human A431 and mouse C2C12 treated with calyculin A. This reactivity is removed with lambda phosphatase treatment.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. This protein also has a role in centrosomal homeostasis by being an activator and substrate of the Aurora A kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-p41-ARCb (Thr21)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene ARPC1B
Gene Accession No. O15143, O88656, Q9WV32
Gene Alias 41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs
Gene Symbols ARPC1B
Host Species Rabbit
Immunogen Phospho-Arpc1b/p41-Arc (Thr-21) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding threonine 21 in human Arpc1b. This peptide sequence is highly conserved in rat, mouse, chicken, and fish Arpc1b, and has some homology to the conserved site in isoform Arpc1a.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10095, 11867, 54227
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.