Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-P53 (Ser37) Polyclonal Antibody, Cyanine5, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3706RCY5
This item is not returnable.
View return policy
Description
Phospho-P53 (Ser37) Polyclonal antibody specifically detects Phospho-P53 (Ser37) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)Specifications
| Phospho-P53 (Ser37) | |
| Polyclonal | |
| Cyanine5 | |
| Tp53 | |
| Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
| KLH conjugated synthetic phosphopeptide derived from human P53 around the phosphorylation site of Ser37 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| P04637 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7157 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction