Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-p53 (Ser376) Rabbit anti-Human, Clone: 7C9, Bioss
SDP

Catalog No. 501989625
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9625 Supplier Bioss Supplier No. BSM52181R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

The tumor suppressor protein, p53, is a sequence specific transcription factor that is activated by cellular stress. p53 mediates cell cycle arrest or apoptosis in response to DNA damage or starvation for pyrimidine nucleotides. p53 is up-regulated in response to stress signals and stimulated to activate transcription of specific genes, resulting in expression of p21waf1 and other proteins involved in G1 or G2/M arrest. The structure of p53 comprises an N-terminal transactivation domain, a central DNA-binding domain, an oligomerisation domain, and a C-terminal regulatory domain. There are various phosphorylation sites on p53, of which the phosphorylation at Ser15 is important for p53 activation and stabilization. p53 has been characterized to play a role in blocking the proliferative action of damaged cells and act as an anticancer agent. Phosphorylation of Ser392 in p53 has been shown to associate with the formation of human tumors. In addition, p53 has also been linked to the effects of aging and oxidative stress and an increase in p53 has been linked to deficits in LTP (Long Term Potentiation) in learning and memory. p53 is found in very low levels in normal cells, however, in a variety of transformed cell lines, it is expressed in high amounts, and believed to contribute to transformation and malignancy. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and cause the loss of tumor suppressor activity. Alterations of the TP53 gene occur not only as somatic mutations in human malignancies, but also as germline mutations in some cancer-prone families such as Li-Fraumeni syndrome.

Specifications

Antigen Phospho-p53 (Ser376)
Applications Western Blot
Classification Monoclonal
Clone 7C9
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Tp53
Gene Accession No. P04637
Gene Alias 53BP1; Antigen NY-CO-13; bbl; BCC7; bfy; bhy; BOS_18908; Cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; m53BP1; murine p53-binding protein; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; P53; p53 cellular tumor antigen; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; p53-binding protein 1; p53BP1; Phosphoprotein p53; tp53; tp53.L; TP53-binding protein 1; Tp53bp1; transformation related protein 53; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; transformation-related protein 53; Trp248; Trp53; Trp53bp1; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; Tumor suppressor p53; tumor suppressor p53-binding protein 1; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Gene Symbols Tp53
Host Species Rabbit
Immunogen Synthetic peptide derived from human p53 (S376), around 350-400aa (phospho S376).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.