Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-p62 (Ser24) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABC204725MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC204725MI 25 μL
ABC204710MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABC204725MI Supplier MilliporeSigma Supplier No. ABC204725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho-p62 (Ser24) Polyclonal antibody specifically detects phospho-p62 (Ser24) in Human,Mouse,Feline,Rhesus Macaque,Rat samples. It is validated for Dot Blot, ELISA, Peptide Array, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-p62 (Ser24)
Applications Dot Blot, ELISA, Peptide Array, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Accession No. NP_003891
Gene Alias Sequestosome-1;EBI3-associated protein of 60 kDa;EBIAP;p60;Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa;Ubiquitin-binding protein p62
Gene Symbols SQSTM1, ORCA, OSIL
Host Species Rabbit
Immunogen Linear peptide corresponding to 8 amino acids surrounding phosphoserine 24 in the N-terminal region of human p62.
Molecular Weight of Antigen ∼62 kDa observed; 47.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects Sequestosome-1 (p62) phosphorylated on serine 24.
Target Species Human, Mouse, Feline, Rhesus Macaque, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.