Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-p62 (Ser403) Rat anti-Mouse, Human, Clone: 4F6, MilliporeSigma™

Catalog No. MABC186IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC186IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC186IMI Supplier MilliporeSigma Supplier No. MABC186I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect p62/SQSTM1 (Ser403) using this rat monoclonal Anti-phospho-p62 (Ser403) antibody, clone 4F6. Validated for use in Immunohistochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-p62 (Ser403)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 4F6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SQSTM1,ORCA,OSIL
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to a sequence surrounding pSer403 of human phospho p62/SQSTM1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_003891.1
Test Specificity Clone 4F6 recognizes p62/SQSTM1 phosphorylated at Ser403 in human and mouse.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.