Learn More
Invitrogen™ Phospho-p73 (Thr86) Polyclonal Antibody

Description
Phospho-p73 (Thr86) Polyclonal Antibody detects endogenous levels of p73 only when phosphorylated at Thr86.
Specifications
Specifications
| Antigen | Phospho-p73 (Thr86) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TP73 |
| Gene Accession No. | O15350 |
| Gene Alias | deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 |
| Gene Symbols | TP73 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.