Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PAK3 (Ser186) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105433 Supplier Thermo Scientific Supplier No. PA5105433
Add to Cart
Add to Cart

Phospho-PAK3 (Ser186) Polyclonal Antibody

Phospho-PAK3 (Ser186) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PAK3 (Ser186)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PAK3
Gene Accession No. O75914, Q61036, Q62829
Gene Alias 2.7.11.1; adriamycin resistance-associated; ARA; beta-PAK; bPAK; CDC42/RAC effector kinase PAK-B; CDKN1A; hPAK3; MRX30; MRX47; oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 3; p21 (RAC1) activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 3; p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p21-activated kinase 3; p65-PAK; PAK beta; Pak3; Pak-3; PAK3beta; Pak65alpha; Pak65beta; Pakb; Serine/threonine-protein kinase PAK 3; Stk4
Gene Symbols PAK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PAK3(Accession O75914), corresponding to amino acid residues around phosphorylated Ser186.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18481, 29433, 5063
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.