Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PAK4/6/7 (Ser474) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564495
Description
Phospho-PAK4/PAK5/PAK6 (Ser474) Polyclonal Antibody detects endogenous levels of PAK4/PAK5/PAK6 only when phosphorylated at Ser474.
PAK4 is highly expressed in prostate, testis and colon. PAK4 interacts tightly with GTP-bound but not GDP-bound CDC42 and weakly with RAC. PAK4 phosphorylates and autophosphorylates and also activates the JNK pathway. PAK5, a member of the PAK family of protein kinases contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK5 preferentially binds to CDC42 in the presence of GTP and the CRIB motif is essential for this interaction. PAK6 protein cotranslocates into the nucleus with androgen receptor, which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development, in response to androgen. PAK6 was initially identified as an androgen receptor in a yeast two hybrid screen and was found to be highly expressed in testis and prostate tissues. Later experiments have shown it to be activated by MAP kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.
Specifications
Phospho-PAK4/6/7 (Ser474) | |
Polyclonal | |
Unconjugated | |
Pak4 | |
2900083L08Rik; 4732456M09; 5730488L07Rik; 6430627N20; AW555722; KIAA1142; KIAA1264; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (CDKN1A)-activated kinase 6; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 4; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 6; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 6; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 4; p21(CDKN1A)-activated kinase 6; p21(CDKN1A)-activated kinase 7; p21-activated kinase 4; p21-activated kinase 5; p21-activated kinase 6; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK 4/5/6; PAK 4/6/7; PAK4; PAK-4; PAK4/5/6; PAK5; PAK-5; PAK6; PAK-6; Pak7; PAK-7; protein kinase PAK5; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 6; serine/threonine-protein kinase PAK 6-like; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10298, 214230, 241656, 292756, 296078, 311450, 56924, 57144, 70584 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
D4A280, O96013, Q3ULB5, Q8BTW9, Q8C015, Q9NQU5, Q9P286 | |
Pak4, PAK5, PAK6, Pak7 | |
A synthesized peptide derived from human PAK4/5/6 (Phospho-Ser474). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction