Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-Pannexin 1 (Tyr308) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN1680
Description
Anti-phospho-Pannexin 1 (Tyr308) Antibody is a highly specific rabbit polyclonal antibody that targets Pannexin 1 and has been tested in Immunohistochemistry and Western Blotting.
Specifications
phospho-Pannexin 1 (Tyr308) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antibody specifically detects pannexin 1 phosphorylated on tyrosine 308 in rat. Also predicted to react with human and chimpanzee. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG |
Immunohistochemistry, Western Blot | |
Please refer to lot specific datasheet. | |
Panx1,Px1 | |
A linear peptide corresponding to 13 amino acids surrounding phosphotyrosine 308 of rat pannexin-1. | |
100 μL | |
Neuroscience | |
NP_955429 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction