Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PAX2 (Ser393) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585758
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Sheep (100%). Positive Control: HCT116, HCT116 nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The pax genes are a family of transcription factors that are active in specific tissues during early embryonic development. Pax family members possess a DNA-binding domain encoded by the paired box. Because paired domain containing genes encode transcription factors, they are capable of executing a genetic program. Several pax genes have also been associated with developmental mutations including: pax-3, which is associated with Waardenburg syndrome, pax-6, which is associated with Aniridia, and pax-2, which is associated with Wilms tumor. During embryogenesis, Pax-2 is expressed in the developing kidney. In particular, the pax-2 gene is expressed in condensing metanephric mesenchyme and in early epithelial structures derived from mesenchyme; however, pax-2 mRNA and protein levels are rapidly down regulated as the tubular epithelium matures. Although Pax-2 is down regulated during renal epithelium maturation, Pax-2 expression persists in the undifferentiated epithelium of Wilms' tumors. Persistent expression of Pax-2 in Wilms' tumors occurs frequently and correlates with the proliferation of poorly differentiated epithelial cells in these tumors. Interestingly, expression of the Wilms' tumor suppresser protein, WT1, coincides with down-regulation of the pax-2 gene; thus, suggesting that WT1 can directly repress pax-2 transcription.
Specifications
Phospho-PAX2 (Ser393) | |
Polyclonal | |
Unconjugated | |
PAX2 | |
FSGS7; Opdc; optic disc coloboma; paired box 2; paired box gene 2; paired box homeotic gene 2; paired box protein Pax-2; PAPRS; PAX2; Pax-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18504, 5076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.57 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P32114, Q02962 | |
PAX2 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse PAX2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction