Learn More
Invitrogen™ Phospho-PAX7 (Ser203) Polyclonal Antibody

Description
Antibody detects endogenous levels of PAX7 only when phosphorylated at Ser203.
Specifications
Specifications
| Antigen | Phospho-PAX7 (Ser203) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PAX7 |
| Gene Accession No. | P23759, P47239 |
| Gene Alias | FLJ37460; HUP1; paired box 7; paired box gene 7; paired box homeotic gene 7; paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; PAX7; Pax-7; PAX7 transcriptional factor; PAX7A; PAX7B; RGD1564360; RMS2 |
| Gene Symbols | PAX7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.