Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Paxillin (Ser83) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143736 Supplier Invitrogen™ Supplier No. PA5143736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a 68kDa* protein corresponding to the molecular mass of phosphorylated paxillin on SDS-PAGE immunoblots of NGF treated rat PC12 cells, as well as calyculin A treated human A431 cells and mouse macrophages.

Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Paxillin (Ser83)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene PXN
Gene Accession No. Q8VI36
Gene Alias adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12
Gene Symbols PXN
Host Species Rabbit
Immunogen Phospho-Paxillin (Ser-83) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 83 of mouse paxillin. This sequence is highly conserved in rat and human paxillin and is also found in all isoforms (a, b, g) of paxillin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19303
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.