Learn More
Invitrogen™ Phospho-Paxillin (Tyr118) Monoclonal Antibody (GT162)

Description
Positive Control: A431 (1 mM pervanadate, 37 degrees C treatment for 30 min), HeLa Predicted Reactivity: Mouse (100%), Rat (100%), Chicken (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-Paxillin (Tyr118) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT162 |
| Concentration | 0.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | PXN |
| Gene Accession No. | P49023 |
| Gene Alias | adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.