Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PDE11A (Ser117) Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208660
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8660 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8660 Supplier Fabgennix Inc Supplier No. PPD11A140AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-PDE11A (Ser117) Polyclonal antibody specifically detects Phospho-PDE11A (Ser117) in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Phospho-PDE11A (Ser117)
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene PDE11A
Gene Accession No. P0C1Q2, Q8VID6, Q9HCR9
Gene Alias 6330414F14Rik; A630086N24Rik; cAMP and cGMP cyclic nucleotide phosphodiesterase 11A; cAMP and cGMP phosphodiesterase 11A; Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A; FLJ23693; Gm350; MGC133355; MGC133356; Pde11a; PDE11A1; phosphodiesterase 11A; PPNAD2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 75-125 on PDE11A protein. Phosphorylated Ser 117.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140928, 241489, 50940
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.