Learn More
Invitrogen™ Phospho-PDHA1 (Ser293) Recombinant Rabbit Monoclonal Antibody (6E3J0)

Description
Phospho-PDHA1 (Ser293) Recombinant Monoclonal Antibody for ELISA, IP, Western Blot
Specifications
Specifications
| Antigen | Phospho-PDHA1 (Ser293) |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 6E3J0 |
| Concentration | 0.58 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | PDHA1 |
| Gene Accession No. | P08559, P26284, P35486 |
| Gene Alias | PDHA; PDHA1; Pdha-1; PDHAD; PDHCE1A; PDHE1-A type I; PHE1A; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1 component alpha subunit; pyruvate dehydrogenase E1 component alpha subunit precursor; pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1 subunit alpha 1; pyruvate dehydrogenase E1alpha subunit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.