Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PDLIM5 (Tyr251) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540209
Description
Phospho-PDLIM5 (Tyr251) Polyclonal Antibody for Western Blot, ELISA
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.
Specifications
Phospho-PDLIM5 (Tyr251) | |
Polyclonal | |
Unconjugated | |
PDLIM5 | |
1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10611 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96HC4 | |
PDLIM5 | |
Synthesized peptide derived from human PDLIM5 around the phosphorylation site of Y251. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction