Learn More
Invitrogen™ Phospho-PDPK1 (Ser241) Polyclonal Antibody

Description
Positive test controls include: NIH/3T3. The target is usually found in the following locations: Cell junction, Cell membrane, Cytoplasm, Nucleus, Peripheral membrane protein, focal adhesion. Immunogen sequence: ANSFV.
Specifications
Specifications
| Antigen | Phospho-PDPK1 (Ser241) |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.39 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Pdpk1 |
| Gene Accession No. | O15530, O55173, Q9Z2A0 |
| Gene Alias | 3-phosphoinositide dependent protein kinase 1; 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; HGNC:8809; hPDK1; isoenzyme 1; kinase isoenzyme 1; lipoamide; mPDK1; OTTHUMP00000174525; OTTHUMP00000205076; PDK1; PDPK1; PDPK2; PDPK2P; Pkb kinase; PkB kinase like gene 1; PkB-like 1; PRO0461; Protein kinase B kinase |
| Gene Symbols | Pdpk1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.