Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PEA15 (Ser104) Polyclonal Antibody
Phospho-PEA15 (Ser104) Polyclonal Antibody
Supplier: Thermo Scientific PA597398
Description
Phospho-PEA15 (Ser104) Polyclonal Antibody for Western Blot

Specifications
Phospho-PEA15 (Ser104) | |
Polyclonal | |
Unconjugated | |
PEA15 | |
15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18611, 364052, 8682 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q15121, Q5U318, Q62048 | |
PEA15, Pea15a | |
A synthetic phosphorylated peptide around S104 of human PEA15 (NP_0037591). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction