Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PFKFB2 (Ser483) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105128 Supplier Invitrogen™ Supplier No. PA5105128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PFKFB2 only when phosphorylated at Ser483.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The AGC kinase group consists of 63 kinases including the cyclic nucleotide-regulated protein kinase (PKA & PKG) family, the diacylglycerol-activated/phospholipid-dependent protein kinase C (PKC) family, the related to PKA and PKC (RAC/Akt) protein kinase family, the kinases that phosphorylate G protein-coupled receptors family (ARK), and the kinases that phosphorylate ribosomal protein S6 family (RSK).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PFKFB2 (Ser483)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pfkfb2
Gene Accession No. O60825, P70265, Q9JJH5
Gene Alias 4930568D07Rik; 6PF-2-K/Fru-2,6-P2ase 2; 6PF-2-K/Fru-2,6-P2ase heart-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase /fructose-2,6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2, 6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2; 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2 (heart); 6-phosphofructo-2-kinase/fructose2,6-bisphosphatase isoenzyme 2; fructose 6-P,2-kinase:fructose 2,6-bisphosphatase; fructose 6-phosphate 2-kinase/fructose 2,6-bisphosphatase; fructose 6-phosphate, 2-kinase:fructose 2,6-bisphosphatase; fructose 6-phosphate,2-kinase:fructose 2, 6-bisphosphatase; Fructose-2,6-bisphosphatase; fructose-2,6-bisphosphatase, cardiac isozyme; PFK/FBPase 2; PFK-2 car; PFK-2 cardiac; PFK-2/FBPase-2; PFK-2/FBPase-2 gene B; PFKFB, cardiac; PFKFB2; RH2K
Gene Symbols Pfkfb2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PFKFB2(Accession O60825), corresponding to amino acid residues around phosphorylated Ser483.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18640, 24640, 5208
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.