Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PGC (Ser15) Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA540191

Catalog No. PA540191


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with human based on sequence homology.

Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity, and although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. The amino acid composition of Pepsin C differs from those of pepsinogen and pepsin especially in the content of basic amino acids, glutamic acid, aspartic acid, leucine and isoleucine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-PGC (Ser15)
Polyclonal
Unconjugated
PGC
2210410L06Rik; Gastricsin; pep carboxylase; PEPC; pepcase; pepsin C; pepsinogen C; pepsinogen group II; PG1; Pg-1; Pgc; PGII; preprogastricsin; progastricsin; progastricsin (pepsinogen C); Upg1; Upg-1; Urinary pepsinogen 1
Rabbit
Antigen Affinity Chromatography
RUO
5225
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P20142
PGC
Synthesized peptide derived from human PEPC around phosphorylation site of S15.
100 μL
Primary
Plant
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.