Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PGK1 (Tyr324) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106227
Description
Antibody detects endogenous levels of PGK1 only when phosphorylated at Y324.
Also known as ATP:3-phosphoglycerate 1-phosphotransferase showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.
Specifications
| Phospho-PGK1 (Tyr324) | |
| Polyclonal | |
| Unconjugated | |
| PGK1 | |
| cell migration-inducing gene 10 protein; epididymis secretory sperm binding protein Li 68p; HEL-S-68p; MIG10; OK/SW-cl.110; Pgk; Pgk1; Pgk-1; PGKA; Phosphoglycerate kinase; phosphoglycerate kinase 1; Primer recognition protein 2; PRP 2; YCR012W; YCR12W | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 18655, 24644, 5230 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00558, P09411, P16617 | |
| PGK1 | |
| A synthesized peptide derived from human PGK1(Accession P00558), corresponding to amino acid residues around phosphorylated Tyr324. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction