Learn More
Invitrogen™ Phospho-PI3K p85/p55 (Tyr458, Tyr199) Recombinant Rabbit Monoclonal Antibody (PI3KY458-1A11), PE
Rabbit Recombinant Monoclonal Antibody
$245.00 - $579.50
Specifications
Antigen | Phospho-PI3K p85/p55 (Tyr458, Tyr199) |
---|---|
Clone | PI3KY458-1A11 |
Content And Storage | 4°C |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA528027
|
Invitrogen™
MA528027 |
100 Tests |
Each for $579.50
|
|
|||||
PIMA536954
|
Invitrogen™
MA536954 |
10 Tests |
Each for $245.00
|
|
|||||
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phosphoinositide 3-kinase (PI3K, phospho PI3K) Binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phospho PI3K is necessary for the insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. During insulin stimulation, it also binds to IRS-1. Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration, and cell survival.Specifications
Phospho-PI3K p85/p55 (Tyr458, Tyr199) | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG κ | |
Human, Mouse | |
PIK3R1 | |
AA414954; AGM7; GRB1; IMD36; mitochondrial poly(ADP-ribose) glycohydrolase; MPPH; MPPH1; OTTHUMP00000009783; OTTHUMP00000009784; p50alpha; p55; p55alpha; p55-GAMMA; p55PIK; p85; p85alpha; p85-ALPHA; P85B; p85beta; p85-BETA; PARG; PARG99; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; phosphatidylinositol 3-kinase regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphoinositide 3-kinase p85 (other splicing variants: p55 and p50); Phosphoinositide 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit 1; phosphoinositide-3-kinase regulatory subunit 2; phosphoinositide-3-kinase regulatory subunit 3; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3K; PI3K regulatory subunit alpha; PI3K regulatory subunit beta; PI3K regulatory subunit gamma; PI3KA; PI3-kinase p85 subunit alpha; PI3-kinase p85 subunit beta; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit beta; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; PI3-kinase subunit p85-beta; Pik3r1; PIK3R2; Pik3r3; poly (ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform; polypeptide 3; ptdIns-3-kinase p85-alpha; ptdIns-3-kinase p85-beta; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit alpha; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-alpha; ptdIns-3-kinase regulatory subunit p85-beta; RP4-533D7.2 | |
PIK3R1, Pik3r2, PIK3R3 | |
Primary | |
Protein A/G |
PI3KY458-1A11 | |
Flow Cytometry | |
PE | |
Rabbit | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O00459, O08908, P26450, P27986, Q64143, Q92569 | |
18708, 18709, 18710, 5295, 5296, 8503 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr458 of human phospho PI3K p85. | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.