Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PI3KCA (Tyr317) Polyclonal Antibody, Cyanine3, Bioss
SDP

Catalog No. 502279373
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9373 Supplier Bioss Supplier No. BS5570RCY3
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-PI3KCA (Tyr317) Polyclonal antibody specifically detects Phospho-PI3KCA (Tyr317) in Bovine, Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen Phospho-PI3KCA (Tyr317)
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine3
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene PIK3CA
Gene Accession No. P32871, P42336, P42337
Gene Alias 2.7.1.153; 6330412C24Rik; caPI3K; CLOVE; CWS5; EC:2.7.11.1; GRB1; MCAP; MCM; MCMTC; MGC142161; MGC142163; p110; p110alpha; p110-alpha; phophatidylinositol 3-kinase; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3K; PI3K alpha; PI3Kalpha; PI3K-alpha; pi3kca; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; Pik3ca; PIK3R1; pk3ca; ptdIns-3-kinase p110; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; Serine/threonine protein kinase PIK3CA
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human PI3KCA around the phosphorylation site of Tyr317
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18706, 282306, 5290
Target Species Bovine, Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.