Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PKA alpha (Ser338) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564489 Supplier Invitrogen™ Supplier No. PA564489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-PKA alpha (Ser338) Polyclonal Antibody detects endogenous levels of PKA alpha only when phosphorylated at Ser338.

Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKA alpha (Ser338)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Gene PRKACA
Gene Accession No. P05132, P17252, P27791
Gene Alias alternatively spliced; C(s); Calpha; C-alpha-2; C-alpha-S; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; cAMP-dependent protein kinase catalytic subunit Calpha1; Cs-PKA; MGC102831; MGC48865; PKA; PKA C-alpha; PKA catalytic subunit alpha; pkac; Pkaca; PKCA1; PKCD; PPNAD4; PRKACA; protein kinase; protein kinase A; protein kinase A alpha; protein kinase A catalytic subunit; protein kinase cAMP-activated catalytic subunit alpha; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; sperm cAMP-dependent protein kinase catalytic subunit; sperm cAMP-dependent protein kinase catalytic subunit Cs
Gene Symbols PRKACA
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKA alpha CAT (Phospho-Ser338).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18747, 25636, 5566
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.