Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PKC Pan (Ser660) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597368 Supplier Invitrogen™ Supplier No. PA597368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T Immunogen sequence: YVNPQ.

Members of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKC Pan (Ser660)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Prkci
Gene Accession No. F1M7Y5, P05129, P05696, P05771, P09215, P09216, P09217, P16054, P17252, P20444, P23298, P24723, P28867, P41743, P63318, P63319, P68403, P68404, Q02111, Q02156, Q02956, Q04759, Q05513, Q05655, Q62074, Q64617, Q9WTQ0
Gene Alias 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; 2310021H13Rik; 5830406C15Rik; A130035A12Rik; A130082F03Rik; AAG6; aging-associated gene 6; AI098070; AI385711; AI427505; AI875142; ALPS3; aPKClambda; aPKC-lambda/iota; aPKCzeta; atypical protein kinase C; atypical protein kinase C-lambda/iota; AW494342; C80388; CVID9; D14Ertd420e; DXS1179E; MAY1; mKIAA4165; nPKC-delta; nPKC-epsilon; nPKC-eta; nPKC-iota; nPKC-theta; nPKC-zeta; PKC; PKC theta; PKC zeta; PKC[d]; PKC[e]; PKC-0; PKC2; Pkca; PKC-A; PKC-alpha; Pkcb; PKC-B; PKC-beta; Pkcc; PKCD; PKCdelta; Pkce; Pkcea; PKCepsilon; PKCG; PKCgamma; PKC-gamma; Pkch; PKCI; PKCL; PKC-L; PKClambda; Pkcq; PKCtheta; PKC-theta; Pkcz; PKC-ZETA; PRKACA; Prkc; PRKCA; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcd; Prkce; PRKCG; Prkch; PRKCI; PRKCL; PRKC-lambda/iota; Prkcq; PRKCT; Prkcz; protein kinase C alpha; protein kinase C alpha type; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C eta; protein kinase C eta type; protein kinase C gamma; protein kinase C gamma type; protein kinase C iota; protein kinase C iota type; protein kinase C theta; protein kinase C theta type; protein kinase C type I (gamma type); protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, alpha; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C, epsilon; protein kinase C, eta; protein kinase C, gamma; protein kinase C, iota; protein kinase C, lambda; protein kinase C, theta; protein kinase C, zeta; protein kinase C[d]; protein kinase c-alpha; protein kinase C-eta; r14-3-3; R74924; R75156; RATPKCI; SCA14; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; zetaPKC
Gene Symbols Prkca, PRKCB, PRKCD, PRKCE, PRKCG, Prkch, Prkci, PRKCQ, PRKCZ
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S660 of human PKC (pan) βIIPRKCA (NP_0027282).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170538, 18750, 18751, 18752, 18753, 18754, 18755, 18759, 18761, 18762, 24680, 24681, 25023, 25522, 29340, 5578, 5579, 5580, 5581, 5582, 5583, 5584, 5588, 5590, 81749, 84006, 85420
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.