Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PKC Pan (Thr514) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597383
Description
Positive Samples: RAW264.7, C6 Immunogen sequence: YQPYG.
Members of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family members exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.
Specifications
| Phospho-PKC Pan (Thr514) | |
| Polyclonal | |
| Unconjugated | |
| Prkca | |
| 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; 2310021H13Rik; 5830406C15Rik; A130035A12Rik; A130082F03Rik; AAG6; aging-associated gene 6; AI098070; AI385711; AI427505; AI875142; ALPS3; aPKClambda; aPKC-lambda/iota; aPKCzeta; atypical protein kinase C; atypical protein kinase C-lambda/iota; AW494342; C80388; CVID9; D14Ertd420e; DXS1179E; MAY1; mKIAA4165; nPKC-delta; nPKC-epsilon; nPKC-eta; nPKC-iota; nPKC-theta; nPKC-zeta; PKC; PKC theta; PKC zeta; PKC[d]; PKC[e]; PKC-0; PKC2; Pkca; PKC-A; PKC-alpha; Pkcb; PKC-B; PKC-beta; Pkcc; PKCD; PKCdelta; Pkce; Pkcea; PKCepsilon; PKCG; PKCgamma; PKC-gamma; Pkch; PKCI; PKCL; PKC-L; PKClambda; Pkcq; PKCtheta; PKC-theta; Pkcz; PKC-ZETA; PRKACA; Prkc; PRKCA; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcd; Prkce; PRKCG; Prkch; PRKCI; PRKCL; PRKC-lambda/iota; Prkcq; PRKCT; Prkcz; protein kinase C alpha; protein kinase C alpha type; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C eta; protein kinase C eta type; protein kinase C gamma; protein kinase C gamma type; protein kinase C iota; protein kinase C iota type; protein kinase C theta; protein kinase C theta type; protein kinase C type I (gamma type); protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, alpha; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C, epsilon; protein kinase C, eta; protein kinase C, gamma; protein kinase C, iota; protein kinase C, lambda; protein kinase C, theta; protein kinase C, zeta; protein kinase C[d]; protein kinase c-alpha; protein kinase C-eta; r14-3-3; R74924; R75156; RATPKCI; SCA14; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; zetaPKC | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170538, 18750, 18751, 18752, 18753, 18754, 18755, 18759, 18761, 18762, 24680, 24681, 25023, 25522, 29340, 5578, 5579, 5580, 5581, 5582, 5583, 5584, 5588, 5590, 81749, 84006, 85420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1M7Y5, P05129, P05696, P05771, P09215, P09216, P09217, P16054, P17252, P20444, P23298, P24723, P28867, P41743, P63318, P63319, P68403, P68404, Q02111, Q02156, Q02956, Q04759, Q05513, Q05655, Q62074, Q64617, Q9WTQ0 | |
| Prkca, PRKCB, PRKCD, PRKCE, PRKCG, Prkch, Prkci, PRKCQ, PRKCZ | |
| A synthetic phosphorylated peptide around T514 of human PKC (pan) (gamma)PRKCA (NP_0027282). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction