Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PKC zeta/lambda (Thr410, Thr403) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517837 Supplier Invitrogen™ Supplier No. PA517837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with endogenous levels of other PKC isoforms.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PKC zeta/lambda (Thr410, Thr403)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PRKCH
Gene Accession No. P09217, P23298, P24723, Q02956, Q05513, Q64617
Gene Alias 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; nPKC-eta; nPKC-zeta; PKC2; Pkch; PKCL; PKC-L; Pkcz; PKC-ZETA; PRKCH; PRKCI; PRKCL; PRKCZ; protein kinase C eta; protein kinase C eta type; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, eta; protein kinase C, zeta; protein kinase C-eta; r14-3-3; R74924; zetaPKC
Gene Symbols PRKCH, PRKCZ
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pThr410 of human PKC zeta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18755, 18762, 25522, 5583, 5590, 81749
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.