Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKMYT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103001 Supplier Invitrogen™ Supplier No. PA5103001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Myt1.

PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant PKMYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKMYT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pkmyt1
Gene Accession No. Q99640, Q9ESG9
Gene Alias 6230424P17; AW209059; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; membrane-associated tyrosine-and threonine-specific cdc2-inhibitory kinase; Myt1; Myt1 kinase; Pkmyt1; pkmyt1.S; PPP1R126; protein kinase, membrane associated tyrosine/threonine 1; protein kinase, membrane associated tyrosine/threonine 1 S homeolog; protein phosphatase 1, regulatory subunit 126; RGD1305434; XELAEV_18047881mg
Gene Symbols Pkmyt1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKMYT1(Accession Q99640), corresponding to amino acid residues S94-P144.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268930, 9088
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.