Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PLCB3 (Ser537) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538389
Description
This gene encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
Specifications
| Phospho-PLCB3 (Ser537) | |
| Polyclonal | |
| Unconjugated | |
| PLCB3 | |
| 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3, PLC beta 3, PLC-beta-3, phosphoinositide phospholipase C-beta-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18797, 29322, 5331 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01970, P51432, Q99JE6 | |
| PLCB3 | |
| A synthetic phosphopeptide derived from human PLC beta 3 around the phosphorylation site of Ser537 (Q-K-SP-L-G) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction