Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PLCg1 (Tyr783) Recombinant Rabbit Monoclonal Antibody (PLCG1Y783-C4), FITC
GREENER_CHOICE

Catalog No. PIMA537028
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537028 10 Tests
PIMA528029 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537028 Supplier Invitrogen™ Supplier No. MA537028
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PLCg1 (Tyr783)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone PLCG1Y783-C4
Conjugate FITC
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene PLCG1
Gene Accession No. P19174, Q62077
Gene Alias 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1; AI894140; Cded; cell differentiation and embryonic development; EC 3.1.4.11; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; NCKAP3; OTTHUMP00000178982; phosphatidylinositol phospholipase C; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-1; phospholipase C gamma 1; phospholipase C, gamma 1; phospholipase C, gamma 1 (formerly subtype 148); phospholipase C-1; phospholipase C-148; Phospholipase C2; phospholipase C-gamma-1; Phospholipase C-II; PLC gamma1; PLC1; Plc-1; PLC148; PLC-148; PLC2I; Plcg1; Plcg-1; PLCgamma1; Plc-gamma1; PLC-gamma-1; PLC-II; PPLCA; triphosphoinositide phosphodiesterase
Gene Symbols PLCG1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr783 of human phospho PLCgamma1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18803, 5335
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.