Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PLK1 (Thr210) Recombinant Rabbit Monoclonal Antibody (PLK1T210-C2), Invitrogen™

Catalog No. PIMA537370
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537370 20 μL
PIMA537124 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537370 Supplier Invitrogen™ Supplier No. MA537370
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PLK1 (Thr210)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone PLK1T210-C2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.10% BSA and 0.02% sodium azide
Gene Plk1
Gene Accession No. P53350
Gene Alias cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg
Gene Symbols Plk1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Thr210 of human phospho PLK1.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5347
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.