Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FXYD1 (Ser68) Monoclonal Antibody (41AT858.235)
GREENER_CHOICE

Catalog No. PIMA511124
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511124 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511124 Supplier Invitrogen™ Supplier No. MA511124
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phospho-FXYD1 (Ser68) Monoclonal Antibody for Western Blot

This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na,K-ATPase. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FXYD1 (Ser68)
Applications Western Blot
Classification Monoclonal
Clone 41AT858.235
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FXYD1
Gene Accession No. O00168, Q9Z239
Gene Alias 0610012C17Rik; 1110006M24Rik; FXYD domain containing ion transport regulator 1; FXYD domain-containing ion transport regulator 1; FXYD1; Phospholemman; PLM; Pml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Symbols FXYD1
Host Species Mouse
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S68 of mouse PLM.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5348, 56188
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.