Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PNK (Ser114, Thr118) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517013 Supplier Invitrogen™ Supplier No. PA517013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5'-hydroxy and 3'-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PNK (Ser114, Thr118)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Pnkp
Gene Accession No. Q96T60
Gene Alias 1810009G08Rik; 2'(3')-polynucleotidase; AOA4; bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; EIEE10; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); MCSZ; phosphatase/kinase; PKNP; PNK; PNKP; Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; polynucleotide kinase 3'- phosphatase; polynucleotide kinase 3'-phosphatase; polynucleotide kinase-3'-phosphatase
Gene Symbols Pnkp
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues around pSer114 and pThr118 of human PNK1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11284
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.