Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PP1 alpha (Thr320) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114625
Description
Antibody detects endogenous levels of PPP1CA only when phosphorylated at Thr320.
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Phospho-PP1 alpha (Thr320) | |
Polyclonal | |
Unconjugated | |
PPP1CA | |
96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
19045, 24668, 5499 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P62136, P62137, P62138 | |
PPP1CA | |
A synthesized peptide derived from human PPP1CA around the phosphorylation site of Thr320. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction