Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PP1 alpha (Thr320) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117212 Supplier Invitrogen™ Supplier No. PA5117212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, NIH/3T3 Immunogen sequence: PITPP.

This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PP1 alpha (Thr320)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PPP1CA
Gene Accession No. P62136, P62137, P62138
Gene Alias 96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155
Gene Symbols PPP1CA
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T320 of human PPP1CA (NP_0026991).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19045, 24668, 5499
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.