Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PP2A alpha (Tyr307) Recombinant Rabbit Monoclonal Antibody (3F11)
GREENER_CHOICE

Catalog No. PIMA533189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533189 Supplier Invitrogen™ Supplier No. MA533189
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-PP2A alpha (Tyr307) Recombinant Monoclonal Antibody for Western Blot, ELISA

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. An additional protein phosphatase catalytic subunit, PPX (also known as PP4) is a putative member of a novel PP family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PP2A alpha (Tyr307)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3F11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP2CA
Gene Accession No. P67775
Gene Alias B55-GAMMA; hypothetical protein; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; PP2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; Pp2a1; PP2A-alpha; PP2Ac; PP2CA; PP2Calpha; Ppp2ca; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A; protein phosphatase-2A-alpha; R75353; replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product
Gene Symbols PPP2CA
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-PPP2CA (Y307).
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5515
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.