Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PPAR delta (Thr256) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105082
Description
Antibody detects endogenous levels of PPARD only when phosphorylated at Thr256.
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) family. PPARs are nuclear hormone receptors that bind peroxisome proliferators and control the size and number of peroxisomes produced by cells. PPARs mediate a variety of biological processes, and may be involved in the development of several chronic diseases, including diabetes, obesity, atherosclerosis, and cancer. This protein is a potent inhibitor of ligand-induced transcription activity of PPAR alpha and PPAR gamma. It may function as an integrator of transcription repression and nuclear receptor signaling. The expression of this gene is found to be elevated in colorectal cancer cells. The elevated expression can be repressed by adenomatosis polyposis coli (APC), a tumor suppressor protein related to APC/beta-catenin signaling pathway. Knockout studies in mice suggested the role of this protein in myelination of the corpus callosum, lipid metabolism, and epidermal cell proliferation.
Specifications
| Phospho-PPAR delta (Thr256) | |
| Polyclonal | |
| Unconjugated | |
| Ppard | |
| FAAR; MGC3931; Nr1c2; NUC1; NUC-1; NUCI; NUCII; nuclear hormone receptor; nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; OTTHUMP00000016256; OTTHUMP00000016257; peroxisome proliferative activated receptor delta; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor beta; peroxisome proliferator activated receptor delta; peroxisome proliferator activator receptor beta; peroxisome proliferator activator receptor delta; peroxisome proliferator activator receptor, delta; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR[b]; PPARB; Pparb/d; PPARbeta; PPAR-beta; Ppard; PPARdelta; PPAR-delta; PPARdelta/beta | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 19015, 25682, 397297, 5467 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35396, Q03181 | |
| Ppard | |
| A synthesized peptide derived from human PPARD(Accession Q03181), corresponding to amino acid residues around phosphorylated Thr256. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction