Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-PPARgamma (Ser112) Rabbit anti-Mouse,Canine,Human,Rat, KHL, Clone: AW504, MilliporeSigma™

Catalog No. 04816I25UMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04816I25UMI 25 μL
04816I100MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04816I25UMI Supplier MilliporeSigma Supplier No. 04816I25UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

phospho-PPARgamma (Ser112) Monoclonal antibody specifically detects phospho-PPARgamma (Ser112) in Mouse,Canine,Human,Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-PPARgamma (Ser112)
Applications Western Blot
Classification Monoclonal
Clone AW504
Conjugate KLH
Gene Alias Peroxisome proliferator-activated receptor gamma;Nuclear receptor subfamily 1 group C member 3
Gene Symbols PPARG, NR1C3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids surrounding phosphoserine 112 from human PPAR gamma, isoform 2.
Molecular Weight of Antigen ∼55 and 58 kDa observed; 57.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone AW504 is a rabbit monoclonal antibody that specifically detects PPAR gamma phosphorylated on serine 112.
Target Species Mouse, Canine, Human, Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Culture Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.