Learn More
Invitrogen™ Phospho-PPP1R2 (Thr72) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 441160G

Description
Phospho-PPP1R2 (Thr72) Polyclonal Antibody for Western Blot, ICC/IF, IHC
Protein phosphatase-1 (PP1) is one of the main eukaryotic serine/threonine phosphatases. The protein encoded by this gene binds to the catalytic subunit of PP1, strongly inhibiting its activity. Ten related pseudogenes have been found throughout the human genome. Several splice variants encoding different isoforms have been found for this gene.
Specifications
Phospho-PPP1R2 (Thr72) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P41236 | |
PPP1R2 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Inhibitor-2 that contains threonine 72. This sequence is conserved in rabbit, dog, zebrafish, chimpanzee, cow, and chicken. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
PPP1R2 | |
0610025N14Rik; 2310007G06Rik; 4930440J04Rik; 5430408E15Rik; D16Ertd248e; Ipp2; IPP-2; phosphatase inhibitor 2; phosphoprotein phosphatase; PPP1R2; protein phosphatase 1 regulatory inhibitor subunit 2; protein phosphatase 1, regulatory (inhibitor) subunit 2; protein phosphatase 1, regulatory subunit 2; protein phosphatase inhibitor 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5504 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.