Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-PRAS40 (T246) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB68901
This item is not returnable.
View return policy
Description
PRAS40 Monoclonal specifically detects PRAS40 in Human samples. It is validated for Western Blot.Specifications
| PRAS40 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 40 kDa, AKT1 substrate 1 (proline-rich), AKT1S1, Lobe, MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 84335 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 1211A | |
| Western Blot 1 ug/mL | |
| Q96B36 | |
| AKT1S1 | |
| Phosphopeptide containing the human PRAS40 T246 site Accession # Q96B36 | |
| 100 μg | |
| Primary | |
| Detects human and mouse PRAS40 when phosphorylated at T246. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction