Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PRK1 (Thr774, Thr816) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501985330
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5330 Supplier Bioss Supplier No. BS3347R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Receptor-like kinase involved in the control of pollen germination and pollen tube polar growth.

Specifications

Antigen Phospho-PRK1 (Thr774, Thr816)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PKN1
Gene Accession No. O08874, P70268, Q16512, Q16513, Q63433, Q8BWW9
Gene Alias 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; DBK; F730027O18Rik; p140 kinase; PAK1; PAK-1; PAK2; Pak-2; Pkn; PKN gamma; Pkn1; Pkn2; PKN-ALPHA; PRK1; PRK2; Prkcl1; PRKCL2; PRO2042; Protease-activated kinase 1; protease-activated kinase 2; Protein kinase C-like 1; Protein kinase C-like 2; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase N2; protein kinase PKN; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; Protein-kinase C-related kinase 2; serine/threonine kinase 3; serine/threonine kinase 7; serine/threonine protein kinase N; Serine/threonine-protein kinase N1; Serine/threonine-protein kinase N2; serine-threonine kinase N; Serine-threonine protein kinase N; Stk3; Stk7
Gene Symbols PKN1, PKN2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human PRK1 around the phosphorylation site of Thr774.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109333, 207122, 29355, 320795, 5585, 5586
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.