Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PRKD1 (Ser742) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986650
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6650 Supplier Bioss Supplier No. BS5562R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKD1 gene encodes a member of the polycystin protein family. The encoded glycoprotein contains a large N-terminal extracellular region, multiple transmembrane domains and a cytoplasmic C-tail. It is an integral membrane protein that functions as a regulator of calcium permeable cation channels and intracellular calcium homoeostasis. It is also involved in cell-cell/matrix interactions and may modulate G-protein-coupled signal-transduction pathways. It plays a role in renal tubular development, and mutations in this gene cause autosomal dominant polycystic kidney disease type 1 (ADPKD1). ADPKD1 is characterized by the growth of fluid-filled cysts that replace normal renal tissue and result in end-stage renal failure. Splice variants encoding different isoforms have been noted for this gene. Also, six pseudogenes, closely linked in a known duplicated region on chromosome 16p, have been described.

Specifications

Antigen Phospho-PRKD1 (Ser742)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PKD1
Gene Accession No. P98161
Gene Alias Autosomal dominant polycystic kidney disease 1 protein; Autosomal dominant polycystic kidney disease 1 protein homolog; mFLJ00285; PBP; PC1; Pc-1; Pkd1; polycystic kidney disease 1 (autosomal dominant); polycystic kidney disease 1 homolog; polycystic kidney disease 1 homolog; polycystin-1; polycystic kidney disease-associated protein; polycystin 1; polycystin 1, transient receptor potential channel interacting; Polycystin-1; transient receptor potential cation channel, subfamily P, member 1; TRPP1
Gene Symbols PKD1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human PRKD1 around the phosphorylation site of Ser742.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5310
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.