Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Profilin 1 (Ser138) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143752 Supplier Invitrogen™ Supplier No. PA5143752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated Profilin (Ser-138) then affinity purified using Profilin (Ser-138) peptide (without carrier). The antibody detects a 15 kDa band corresponding to the apparent molecular mass of Profilin-1 in human Jurkat and A431 cells, as well as rat PC12 cells. This reactivity is decreased by lambda phosphatase treatment, and is blocked by phospho-profilin (Ser-138) blocking peptide.

Profilin is an ubiquitous small (12-15 kDa) phosphoinositides and poly-L-proline binding protein that plays a role in both signal transduction pathways and actin filament dynamics. There are two mammalian profilins with similar biochemical properties but different expression patterns. Profilin I appears to be highly expressed in most tissues except for skeletal muscle. Profilin II is predominantly expressed in brain and at lower levels in skeletal muscle, uterus and kidney. Profilin is a mainly cytosolic protein with higher concentrations in dynamic membrane areas like the leading edge and ruffling membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Profilin 1 (Ser138)
Applications Western Blot
Classification Polyclonal
Concentration ∼0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Pfn1
Gene Accession No. P07737, P62962, P62963
Gene Alias actin binding protein; ALS18; Epididymis tissue protein Li 184a; Pfn; Pfn1; profilin 1; profilin I; Profilin-1
Gene Symbols Pfn1
Host Species Rabbit
Immunogen Phospho-Profilin (Ser138) synthetic peptide (coupled to carrier protein) that includes amino acids surrounding Ser138 in human Profilin-1. This sequence is conserved in rat and mouse Profilin-1. The serine site has some homology to the conserved site (Ser138) in Profilin-2a and Profilin-2b, but the site is not conserved in Profiin-3 and Profilin-4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18643, 5216, 64303
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.