Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Progesterone R/NR3C3 (S294) Antibody, R&D Systems™

Catalog No. PPS018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS018 Supplier R&D Systems Supplier No. PPS018
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Progesterone R/NR3C3 (p ser294)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 608
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol with No Preservative
Gene Alias NR3C3, NR3C3PRNuclear receptor subfamily 3 group C member 3, PGR, progesterone receptor, ProgesteroneR
Gene Symbols PGR
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5241
Target Species Human, Mouse, Rat, Primate
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.