Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-PRKD2 (Ser876) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540221
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
PRKD2 (PKD2) is a member of the protein kinase D serine/threonine protein kinase family. PRKD2 is a component in the signal transduction pathway induced by phorbol esters and the CCK b gastrin receptor. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKC and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.
Specifications
| Phospho-PRKD2 (Ser876) | |
| Polyclonal | |
| Unconjugated | |
| PRKD2 | |
| AI325941; HSPC187; nPKC-D2; PKD2; Prkd2; protein kinase D2; RGD1308054; serine/threonine-protein kinase D2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 101540, 25865, 292658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| Q5XIS9, Q8BZ03, Q9BZL6 | |
| PRKD2 | |
| Synthesized peptide derived from human PKD2 around the phosphorylation site of S876. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction