Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PSEN2 (Ser330) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105831 Supplier Invitrogen™ Supplier No. PA5105831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Presenilin 2 only when phosphorylated at Ser330.

Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1 or PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor such that, they either directly regulate gamma-secretase activity, or themselves act are protease enzymes. Two alternatively spliced transcript variants encoding different isoforms of PSEN2 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PSEN2 (Ser330)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PSEN2
Gene Accession No. O88777, P49810, Q61144
Gene Alias AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2
Gene Symbols PSEN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSEN2(Accession P49810), corresponding to amino acid residues around phosphorylated Ser330.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19165, 5664, 81751
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.