Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PSMC5 (Ser120) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105192 Supplier Invitrogen™ Supplier No. PA5105192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PSMC5 only when phosphorylated at S120.

Human SUG1 (also known as p45, Rpt6 and Cim3) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP-dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PSMC5 (Ser120)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Psmc5
Gene Accession No. P62195, P62196, P62198
Gene Alias 26S protease regulatory subunit 8; 26S proteasome AAA-ATPase subunit RPT6; 26S proteasome regulatory subunit 8; for proteasomal ATPase (SUG1); mSUG1; MSUG1 protein; p45; p45/SUG; peptidase (prosome, macropain) 26S subunit, ATPase 5; protease (prosome, macropain) 26S subunit, ATPase 5; proteasomal ATPase (SUG1); proteasome (prosome, macropain) 26S subunit, ATPase, 5; proteasome 26S ATPase subunit 5; proteasome 26S subunit ATPase 5; proteasome 26S subunit, ATPase 5; proteasome p45/SUG; proteasome subunit p45; Psmc5; S8; SUG1; SUG-1; Tat-binding protein homolog 10; TBP10; testicular tissue protein Li 149; thy; thyroid hormone receptor-interacting protein 1; thyroid receptor interactor 1; TRIP1
Gene Symbols Psmc5
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSMC5(Accession P62195), corresponding to amino acid residues around phosphorylated Ser120.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19184, 5705, 81827
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.