Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PTPN7 (Ser246) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986645
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6645 Supplier Bioss Supplier No. BS5557R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP was shown to be involved in the regulation of T cell antigen receptor signaling, which was thought to function through dephosphorylating the molecules related to MAP kinase pathway. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen Phospho-PTPN7 (Ser246)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTPN7
Gene Accession No. P35236
Gene Alias BOS_16142; BPTP-4; C920001D21Rik; dual specificity phosphatase 1; Hematopoietic protein-tyrosine phosphatase; HEPTP; Lcptp; LC-PTP; LPTP; protein tyrosine phosphatase, non-receptor type 7; Protein-tyrosine phosphatase LC-PTP; protein-tyrosine phosphatase, non-receptor type 7; protein-tyrosine phosphatase, nonreceptor-type, stress induced; PTPN7; PTPNI; Tyrosine-protein phosphatase non-receptor type 7
Gene Symbols PTPN7
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from mouse PTPN7 around the phosphorylation site of Ser246.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5778
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.